Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000468085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000468085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000468085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000468085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000468085.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000477026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000477026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000477026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000477026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000494642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000494642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
ENST00000494642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000366899.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000468085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000468085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000468085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000468085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000468085.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000477026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000477026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000477026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000477026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000494642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000494642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
ENST00000494642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NM_007207.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111939.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017420.hg.1 |
NR_111940.1 |
Refseq |