Assay Details
| Gene Symbol: ZBTB8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_111980.1 | 2 - 3 | 502 | 118 | ||
| GenBank mRNA | HY034825.1 | - | 2 - 3 | 184 | 118 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and BTB domain containing 8B | ZBTB8B | ZNF916B | -65.6kb | Sense | Chr. 1 - 32465057 - 32487859 on NCBI Build GRCh38 |
| Downstream | zinc finger and BTB domain containing 8 opposite strand | ZBTB8OS |
ARCH, ARCH2 |
25.6kb | Anti-sense | Chr. 1 - 32620820 - 32651013 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237639.hg.1 | TC0100018209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100237636.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237636.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237636.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237636.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237636.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237636.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237636.hg.1 | TC0100018209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237638.hg.1 | TC0100018209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237649.hg.1 | TC0100018209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100237635.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237635.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237635.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237635.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237635.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237635.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237635.hg.1 | TC0100018209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237641.hg.1 | TC0100018209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018209.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237646.hg.1 | TC0100018209.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | ENST00000316459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | ENST00000373510.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | NM_001040441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | NM_001291496.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | NR_111980.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100237648.hg.1 | TC0100018209.hg.1 | Clariom D TC |

ZBTB8A
Genomic Map