Assay Details
| Gene Symbol: LOC101927560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_119374.1 | 2 - 3 | 259 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1362 | LINC01362 | -803.9kb | Anti-sense | Chr. 1 - 82903183 - 82997971 on NCBI Build GRCh38 | |
| Downstream | tubulin tyrosine ligase like 7 | TTLL7 | 19.4kb | Sense | Chr. 1 - 83865021 - 83999340 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927560
Genomic Map