Assay Details
| Gene Symbol: GALNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120373.1 | 1 - 2 | 215 | 128 | ||
| GenBank mRNA | BF727269.1 | - | 1 - 2 | 149 | 128 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927532 | LOC101927532 | -60.0kb | Sense | Chr. 1 - 230002717 - 230007162 on NCBI Build GRCh38 | |
| Downstream | piggyBac transposable element derived 5 | PGBD5 | 229.6kb | Anti-sense | Chr. 1 - 230321646 - 230425928 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
immunoglobulin biosynthetic process
protein O-linked glycosylation
O-glycan processing
protein O-linked glycosylation via serine
protein O-linked glycosylation via threonine
polypeptide N-acetylgalactosaminyltransferase activity
protein binding
manganese ion binding
carbohydrate binding
immunoglobulin biosynthetic process
protein O-linked glycosylation
O-glycan processing
protein O-linked glycosylation via serine
protein O-linked glycosylation via threonine
polypeptide N-acetylgalactosaminyltransferase activity
protein binding
manganese ion binding
carbohydrate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187954.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187954.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187954.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187954.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187954.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187954.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187954.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187954.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187954.hg.1 | TC0100011920.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100187955.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187955.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187955.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187955.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187955.hg.1 | TC0100011920.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000485438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000488903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000492568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | ENST00000494106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_001291866.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011920.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187952.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187952.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187952.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187952.hg.1 | ENST00000366672.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100187952.hg.1 | NM_004481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187952.hg.1 | NR_120373.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100187952.hg.1 | TC0100011920.hg.1 | Clariom D TC |

GALNT2
Genomic Map