Assay Details
| Gene Symbol: ENOX1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120399.1 | 2 - 3 | 144 | 103 | ||
| GenBank mRNA | CR739820.1 | - | 2 - 3 | 144 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 400 | LINC00400 | -295.8kb | Sense | Chr. 13 - 43113165 - 43159466 on NCBI Build GRCh38 | |
| Downstream | coiled-coil domain containing 122 | CCDC122 | 365.5kb | Anti-sense | Chr. 13 - 43821959 - 43879690 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ENOX1-AS2
Genomic Map