Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007009.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147608.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147608.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147608.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147608.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147608.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147608.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147608.hg.1 |
TC1300007009.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300147607.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147607.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147607.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147607.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147607.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147607.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147607.hg.1 |
TC1300007009.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300147609.hg.1 |
ENST00000434273.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147609.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147609.hg.1 |
ENST00000434273.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300147609.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300147609.hg.1 |
TC1300007009.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
ENST00000434273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
NR_120399.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007009.hg.1 |
NR_120399.1 |
Refseq |