Assay Details
| Gene Symbol: LMO7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120410.1 | 1 - 2 | 168 | 59 | ||
| GenBank mRNA | AI816891.1 | - | 1 - 2 | 168 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin C-terminal hydrolase L3 | UCHL3 | UCH-L3 | -16.2kb | Anti-sense | Chr. 13 - 75549480 - 75606020 on NCBI Build GRCh38 |
| Downstream | LMO7 downstream neighbor | LMO7DN | C13orf45 | 234.6kb | Anti-sense | Chr. 13 - 75869019 - 75883888 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009274.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300162271.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300162271.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300162271.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300162271.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300162271.hg.1 | TC1300009274.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300162272.hg.1 | TC1300009274.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | ENST00000568735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120410.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120411.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120412.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009274.hg.1 | NR_120412.1 | Refseq |

LMO7-AS1
Genomic Map