| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008431.hg.1 |
ENST00000444744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008431.hg.1 |
ENST00000444744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008431.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008431.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008431.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008431.hg.1 |
ENST00000444744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008431.hg.1 |
ENST00000444744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008431.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008431.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008431.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156837.hg.1 |
ENST00000444744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156837.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156837.hg.1 |
ENST00000444744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156837.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156837.hg.1 |
TC1300008431.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300156839.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156839.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156839.hg.1 |
TC1300008431.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300156838.hg.1 |
ENST00000444744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156838.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156838.hg.1 |
ENST00000444744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156838.hg.1 |
NR_120429.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156838.hg.1 |
TC1300008431.hg.1 |
Clariom D TC |