Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200185754.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185754.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200185754.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185754.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200185754.hg.1 |
TC1200011000.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200185759.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185759.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185759.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200185759.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185759.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200185759.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200185759.hg.1 |
TC1200011000.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000550678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
ENST00000552201.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011000.hg.1 |
NR_120452.1 |
Refseq |