Assay Details
| Gene Symbol: LINC01481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120460.1 | 4 - 5 | 339 | 82 | ||
| GenBank mRNA | HY013176.1 | - | 4 - 5 | 339 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myelin regulatory factor-like | MYRFL |
C12orf15, C12orf28, bcm1377 |
-263.5kb | Anti-sense | Chr. 12 - 69824518 - 69958726 on NCBI Build GRCh38 |
| Downstream | CCR4-NOT transcription complex subunit 2 | CNOT2 |
CDC36, HSPC131, NOT2, NOT2H |
17.6kb | Anti-sense | Chr. 12 - 70242993 - 70354993 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000552998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000552998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187053.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187053.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187053.hg.1 | TC1200011230.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200187057.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187057.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187057.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187057.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200187057.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187057.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187057.hg.1 | TC1200011230.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200187052.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187052.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200187052.hg.1 | TC1200011230.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000549651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000550216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000552998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000552998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | ENST00000553135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120460.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011230.hg.1 | NR_120461.1 | Refseq |

LINC01481
Genomic Map