Assay Details
| Gene Symbol: LINC01498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120483.1 | 2 - 3 | 321 | 140 | ||
| GenBank mRNA | DB089475.1 | - | 2 - 3 | 321 | 140 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chemerin chemokine-like receptor 1 | CMKLR1 |
CHEMERINR, ChemR23, DEZ, RVER1 |
-122.3kb | Sense | Chr. 12 - 108288044 - 108339348 on NCBI Build GRCh38 |
| Downstream | FIC domain containing | FICD |
HIP13, HYPE, UNQ3041 |
42.0kb | Anti-sense | Chr. 12 - 108515274 - 108519603 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | NR_120483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | NR_120483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | NR_120483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011842.hg.1 | NR_120483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191711.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191711.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191711.hg.1 | ENST00000551889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191711.hg.1 | NR_120483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191711.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191711.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191711.hg.1 | ENST00000551889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191711.hg.1 | NR_120483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191711.hg.1 | TC1200011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000502160.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | ENST00000551889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | NR_120483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | NR_120483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | NR_120483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011842.hg.1 | NR_120483.1 | Refseq |

LINC01498
Genomic Map