Assay Details
Gene Symbol: LOC102724050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120486.1 | 2 - 3 | 317 | 67 | ||
GenBank mRNA | DA746474.1 | - | 2 - 3 | 216 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 385A | ZNF385A |
HZF, RZF, ZFP385, ZNF385 |
-16.1kb | Anti-sense | Chr. 12 - 54369129 - 54391609 on NCBI Build GRCh38 |
Downstream | gametocyte specific factor 1 | GTSF1 | FAM112B | 46.2kb | Anti-sense | Chr. 12 - 54455952 - 54473602 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155781.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155781.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155781.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155781.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155781.hg.1 | TC1200007763.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200155784.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200155784.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155784.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200155784.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155784.hg.1 | TC1200007763.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200155782.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200155782.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155782.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155782.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200155782.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155782.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155782.hg.1 | TC1200007763.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155783.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200155783.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155783.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155783.hg.1 | ENST00000552785.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200155783.hg.1 | NR_120486.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155783.hg.1 | NR_120487.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155783.hg.1 | TC1200007763.hg.1 | Clariom D TC |
