Assay Details
| Gene Symbol: LOC102724050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120487.1 | 2 - 3 | 322 | 79 | ||
| GenBank mRNA | DA746386.1 | - | 2 - 3 | 324 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 385A | ZNF385A |
HZF, RZF, ZFP385, ZNF385 |
-16.1kb | Anti-sense | Chr. 12 - 54369129 - 54391609 on NCBI Build GRCh38 |
| Downstream | gametocyte specific factor 1 | GTSF1 | FAM112B | 28.0kb | Anti-sense | Chr. 12 - 54455952 - 54473602 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155802.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155802.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155802.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155802.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155802.hg.1 | TC1200007763.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155787.hg.1 | TC1200007763.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200155801.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200155801.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155801.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155801.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200155801.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155801.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200155801.hg.1 | TC1200007763.hg.1 | Clariom D TC |

LOC102724050
Genomic Map