Assay Details
| Gene Symbol: LINC01488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120542.1 | 2 - 3 | 487 | 93 | ||
| NR_120543.1 | 2 - 3 | 487 | 93 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724265 | LOC102724265 | -56.4kb | Sense | Chr. 11 - 69425671 - 69429621 on NCBI Build GRCh38 | |
| Downstream | cyclin D1 | CCND1 |
BCL1, D11S287E, PRAD1, U21B31 |
152.8kb | Sense | Chr. 11 - 69641105 - 69654474 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LINC01488
Genomic Map