Assay Details
| Gene Symbol: LINC01488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120542.1 | 4 - 5 | 1839 | 80 | ||
| NR_120544.1 | 3 - 4 | 1735 | 80 | |||
| NR_120545.1 | 2 - 3 | 1325 | 80 | |||
| GenBank mRNA | KC136297.1 | - | 3 - 4 | 1940 | 80 | |
| KC136300.1 | - | 4 - 5 | 1839 | 80 | ||
| KC136301.1 | - | 3 - 4 | 1734 | 80 | ||
| KC136302.1 | - | 2 - 3 | 1325 | 80 | ||
| KC136303.1 | - | 3 - 4 | 1746 | 80 | ||
| KC136304.1 | - | 4 - 5 | 1645 | 80 | ||
| KC136305.1 | - | 4 - 5 | 1656 | 80 | ||
| KC136306.1 | - | 3 - 4 | 1940 | 80 | ||
| KC136307.1 | - | 4 - 5 | 1788 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724265 | LOC102724265 | -60.9kb | Sense | Chr. 11 - 69425671 - 69429621 on NCBI Build GRCh38 | |
| Downstream | cyclin D1 | CCND1 |
BCL1, D11S287E, PRAD1, U21B31 |
149.3kb | Sense | Chr. 11 - 69641105 - 69654474 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LINC01488
Genomic Map