| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007327.hg.1 |
ENST00000534756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007327.hg.1 |
ENST00000534756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007327.hg.1 |
ENST00000534756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007327.hg.1 |
NR_120553.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007327.hg.1 |
NR_120553.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007327.hg.1 |
NR_120553.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151721.hg.1 |
ENST00000534756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151721.hg.1 |
NR_120553.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151721.hg.1 |
ENST00000534756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151721.hg.1 |
NR_120553.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151721.hg.1 |
TC1100007327.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100007327.hg.1 |
ENST00000534756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007327.hg.1 |
ENST00000534756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007327.hg.1 |
ENST00000534756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007327.hg.1 |
NR_120553.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007327.hg.1 |
NR_120553.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007327.hg.1 |
NR_120553.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151720.hg.1 |
ENST00000534756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151720.hg.1 |
NR_120553.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151720.hg.1 |
ENST00000534756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100151720.hg.1 |
NR_120553.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100151720.hg.1 |
TC1100007327.hg.1 |
Clariom D TC |