| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000531882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000531882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000531882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
ENST00000531882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
NR_120554.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
NR_120554.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
NR_120554.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
NR_120554.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100010569.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
ENST00000531882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
NR_120554.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
ENST00000531882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
NR_120554.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180188.hg.1 |
TC1100010569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000531882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000531882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000531882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
ENST00000531882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
NR_120554.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
NR_120554.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
NR_120554.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
NR_120554.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100010569.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180183.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180183.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180183.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180183.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180183.hg.1 |
ENST00000529017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100180183.hg.1 |
NR_120555.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100180183.hg.1 |
TC1100010569.hg.1 |
Clariom D TC |