| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524488.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007413.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100007413.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100152431.hg.1 |
ENST00000524488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100152431.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100152431.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100152431.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100152431.hg.1 |
ENST00000524488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100152431.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100152431.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100152431.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100152431.hg.1 |
TC1100007413.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100152430.hg.1 |
ENST00000524488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100152430.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100152430.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100152430.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100152430.hg.1 |
ENST00000524488.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100152430.hg.1 |
ENST00000524565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100152430.hg.1 |
NR_120557.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100152430.hg.1 |
NR_120558.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100152430.hg.1 |
TC1100007413.hg.1 |
Clariom D TC |