| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
ENST00000500502.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
ENST00000500502.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
ENST00000530896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
ENST00000530896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
NR_120571.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
ENST00000500502.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
ENST00000500502.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
ENST00000530896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
ENST00000530896.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
NR_120571.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100190980.hg.1 |
TC1100011782.hg.1 |
Clariom D TC |