Assay Details
| Gene Symbol: LOC101929089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120572.1 | 1 - 2 | 275 | 94 | ||
| GenBank mRNA | BE671460.1 | - | 1 - 2 | 69 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetratricopeptide repeat domain 36 | TTC36 | HBP21 | Anti-sense | Chr. 11 - 118527495 - 118531025 on NCBI Build GRCh38 | |
| Downstream | transmembrane protein 25 | TMEM25 | 313.0 | Anti-sense | Chr. 11 - 118531088 - 118546670 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000556583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000556583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000556583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | ENST00000556583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120574.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100196017.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100196017.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100196017.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100196017.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100196017.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100196017.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100196017.hg.1 | TC1100012487.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000525992.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000528578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000532597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000554407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000556583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000556583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000556583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | ENST00000556583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120572.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120573.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120574.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120576.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012487.hg.1 | NR_120576.1 | Refseq |

LOC101929089
Genomic Map