Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010342.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178536.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178536.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178536.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178536.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178536.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178536.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178536.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178536.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178536.hg.1 |
TC1100010342.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010342.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178538.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178538.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178538.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178538.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178538.hg.1 |
ENST00000532380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178538.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178538.hg.1 |
ENST00000534135.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100178538.hg.1 |
NR_120583.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100178538.hg.1 |
TC1100010342.hg.1 |
Clariom D TC |