Assay Details
Gene Symbol: LINC01499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120584.1 | 2 - 3 | 140 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | leucine rich repeat containing 4C | LRRC4C |
NGL-1, NGL1 |
-274.5kb | Anti-sense | Chr. 11 - 40107066 - 41460212 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101928591 | LOC101928591 | 180.3kb | Anti-sense | Chr. 11 - 42001722 - 42085458 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000530379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | ENST00000530379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | ENST00000530379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | ENST00000530379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100151781.hg.1 | TC1100007349.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000530379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | ENST00000530379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007349.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100151785.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151785.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151785.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151785.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100151785.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151785.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151785.hg.1 | ENST00000529282.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151785.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100151785.hg.1 | TC1100007349.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100151777.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151777.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151777.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100151777.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151777.hg.1 | ENST00000528720.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100151777.hg.1 | NR_120584.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100151777.hg.1 | TC1100007349.hg.1 | Clariom D TC |
