Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000530379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
ENST00000530379.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151790.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151790.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151790.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151790.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151790.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151790.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151790.hg.1 |
TC1100007349.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000528720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000529282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000530379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
ENST00000530379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007349.hg.1 |
NR_120584.1 |
Refseq |