Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147630.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147630.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147630.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147630.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147630.hg.1 |
TC1100006793.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100147631.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147631.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147631.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147631.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147631.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147631.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147631.hg.1 |
TC1100006793.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100147628.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147628.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147628.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147628.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147628.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147628.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147628.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147628.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147628.hg.1 |
TC1100006793.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100147633.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147633.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147633.hg.1 |
TC1100006793.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100147627.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147627.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147627.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147627.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147627.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100147627.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100147627.hg.1 |
TC1100006793.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000527725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000532930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
ENST00000533843.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006793.hg.1 |
NR_120590.1 |
Refseq |