| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000178995.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178995.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000178995.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000178995.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000178995.hg.1 |
TC1000011587.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000548010.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
ENST00000552096.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011587.hg.1 |
NR_120618.1 |
Refseq |