Assay Details
| Gene Symbol: FAM53B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120630.1 | 2 - 3 | 440 | 105 | ||
| GenBank mRNA | AW003106.1 | - | 1 - 2 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phospholysine phosphohistidine inorganic pyrophosphate phosphatase | LHPP | HDHD2B | -93.7kb | Sense | Chr. 10 - 124461772 - 124614141 on NCBI Build GRCh38 |
| Downstream | methyltransferase like 10 | METTL10 | C10orf138 | 43.1kb | Anti-sense | Chr. 10 - 124757831 - 124791941 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

FAM53B-AS1
Genomic Map