Assay Details
| Gene Symbol: LINC01517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120652.1 | 1 - 2 | 111 | 89 | ||
| GenBank mRNA | HY052409.1 | - | 1 - 2 | 112 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BMP and activin membrane bound inhibitor | BAMBI | NMA | -60.7kb | Sense | Chr. 10 - 28677495 - 28682939 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 837 | LINC00837 | 45.2kb | Anti-sense | Chr. 10 - 28789311 - 28796050 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000456082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000456082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000456082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000456082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000603866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000603866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000604686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000604686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000611112.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000611112.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000611112.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000611112.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000611112.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148672.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148672.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148672.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148672.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148672.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148672.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148672.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148672.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148672.hg.1 | TC1000007135.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000456082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000456082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000456082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000456082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000603866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000603866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000604686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000604686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000611112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000611112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000611112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000611112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000611112.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000616194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007135.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148643.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148643.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148643.hg.1 | TC1000007135.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148678.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148678.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148678.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148678.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148678.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148678.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148678.hg.1 | TC1000007135.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148677.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148677.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148677.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148677.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148677.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148677.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148677.hg.1 | TC1000007135.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148676.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148676.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148676.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148676.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148676.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148676.hg.1 | ENST00000426922.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148676.hg.1 | ENST00000621861.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148676.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148676.hg.1 | TC1000007135.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000148649.hg.1 | ENST00000456082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148649.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148649.hg.1 | ENST00000456082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000148649.hg.1 | NR_120652.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000148649.hg.1 | TC1000007135.hg.1 | Clariom D TC |

LINC01517
Genomic Map