Assay Details
| Gene Symbol: SPATS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120663.1 | 10 - 11 | 1871 | 72 | ||
| GenBank mRNA | AK095582.1 | - | 10 - 11 | 1871 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FGFR1 oncogene partner 2 pseudogene | LOC100335030 | -120.5kb | Sense | Chr. 12 - 49389174 - 49392335 on NCBI Build GRCh38 | |
| Downstream | potassium voltage-gated channel subfamily H member 3 | KCNH3 |
BEC1, ELK2, Kv12.2 |
24.4kb | Sense | Chr. 12 - 49538541 - 49558312 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SPATS2
Genomic Map