| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000440589.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000440589.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000440589.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000440589.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000440589.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000502725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000502725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000502725.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000505481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000505481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000505481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
ENST00000505481.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
NR_120666.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
NR_120666.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
NR_120666.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011240.hg.1 |
NR_120666.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000176554.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000176554.hg.1 |
ENST00000502725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000176554.hg.1 |
NR_120666.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000176554.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000176554.hg.1 |
ENST00000502725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000176554.hg.1 |
NR_120666.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000176554.hg.1 |
TC1000011240.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000440589.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000440589.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000440589.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000440589.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000440589.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000448936.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000502725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000502725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000502725.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000505481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000505481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000505481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
ENST00000505481.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
NR_120666.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
NR_120666.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
NR_120666.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011240.hg.1 |
NR_120666.1 |
Refseq |