Assay Details
Gene Symbol: GXYLT1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121572.1 | 1 - 2 | 1658 | 66 | ||
GenBank mRNA | DA103059.1 | - | 1 - 2 | 514 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 95 member B1 | FAM95B1 | -19.3kb | Sense | Chr. 9 - 40323571 - 40329220 on NCBI Build GRCh38 | |
Downstream | ribosome biogenesis protein BMS1 homolog | LOC101929959 | 111.7kb | Anti-sense | Chr. 9 - 40568060 - 40579402 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900007309.hg.1 | ENST00000620059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007309.hg.1 | ENST00000620059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170506.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170506.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170506.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170506.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170506.hg.1 | TC0900010281.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900170505.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170505.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170505.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170505.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170505.hg.1 | TC0900010281.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900170507.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170507.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170507.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170507.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170507.hg.1 | TC0900010281.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900170508.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170508.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170508.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170508.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170508.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170508.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170508.hg.1 | TC0900010281.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900170512.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170512.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170512.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170512.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170512.hg.1 | TC0900010281.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000614548.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007308.hg.1 | ENST00000618830.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170510.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170510.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170510.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170510.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170510.hg.1 | TC0900010281.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900170513.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170513.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170513.hg.1 | TC0900010281.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900007309.hg.1 | ENST00000620059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007309.hg.1 | ENST00000620059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170511.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170511.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170511.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170511.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170511.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900170511.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900170511.hg.1 | TC0900010281.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000613576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000615415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | ENST00000616580.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010281.hg.1 | NR_121572.1 | Refseq |
