Assay Details
| Gene Symbol: LOC101929563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121602.1 | 24 - 25 | 3692 | 102 | ||
| GenBank mRNA | CR627240.1 | - | 24 - 25 | 3680 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1239 | LINC01239 | -688.9kb | Anti-sense | Chr. 9 - 22646200 - 22824213 on NCBI Build GRCh38 | |
| Downstream | ELAV like neuron-specific RNA binding protein 2 | ELAVL2 |
HEL-N1, HELN1, HUB |
176.7kb | Sense | Chr. 9 - 23690099 - 23850757 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101929563
Genomic Map