Assay Details
| Gene Symbol: LOC100507291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121609.1 | 3 - 4 | 361 | 100 | ||
| GenBank mRNA | DA364121.1 | - | 3 - 4 | 324 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nicotinamide nucleotide adenylyltransferase 3 | NMNAT3 |
FKSG76, PNAT-3, PNAT3 |
Anti-sense | Chr. 3 - 139560180 - 139678050 on NCBI Build GRCh38 | |
| Downstream | calsyntenin 2 | CLSTN2 |
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma |
357.3kb | Sense | Chr. 3 - 139935185 - 140577397 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000381790.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000381790.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000381790.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000512622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000512622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000512622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000512622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000514729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000514729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000514729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300164272.hg.1 | ENST00000381790.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300164272.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300164272.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300164272.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300164272.hg.1 | ENST00000381790.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300164272.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300164272.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300164272.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300164272.hg.1 | TC0300008971.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000381790.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000381790.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000381790.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000504670.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000507362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000510068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000512622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000512622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000512622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000512622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000514729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000514729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000514729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | ENST00000515247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121608.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121609.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008971.hg.1 | NR_121609.1 | Refseq |

LOC100507291
Genomic Map