| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000504144.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000504144.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000504144.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009012.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161816.hg.1 |
ENST00000504144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161816.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161816.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161816.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161816.hg.1 |
ENST00000504144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161816.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161816.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161816.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161816.hg.1 |
TC0400009012.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
ENST00000504144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
ENST00000504144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161818.hg.1 |
TC0400009012.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000499452.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000504144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000504144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000504144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
ENST00000509332.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
NR_121627.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
NR_121628.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009012.hg.1 |
NR_121628.1 |
Refseq |