Assay Details
| Gene Symbol: LOC100996286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121628.1 | 2 - 3 | 158 | 129 | ||
| GenBank mRNA | CB986668.1 | - | 161 | 129 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100505685 | LOC100505685 | -133.9kb | Sense | Chr. 4 - 151955970 - 151967587 on NCBI Build GRCh38 | |
| Downstream | F-box and WD repeat domain containing 7 | FBXW7 |
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4 |
216.5kb | Anti-sense | Chr. 4 - 152321258 - 152536095 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC100996286
Genomic Map