Assay Details
| Gene Symbol: LOC101928105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121657.1 | 2 - 3 | 305 | 165 | ||
| GenBank mRNA | HY000320.1 | - | 2 - 3 | 306 | 165 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ERICH6 antisense RNA 1 | ERICH6-AS1 | -14.3kb | Anti-sense | Chr. 3 - 150703564 - 150720146 on NCBI Build GRCh38 | |
| Downstream | siah E3 ubiquitin protein ligase 2 | SIAH2 | hSiah2 | 4.6kb | Sense | Chr. 3 - 150741123 - 150763476 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928105
Genomic Map