Assay Details
| Gene Symbol: LINC00824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121672.1 | 4 - 5 | 1328 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1208 | MIR1208 |
MIRN1208, hsa-mir-1208 |
-271.6kb | Anti-sense | Chr. 8 - 128150116 - 128150188 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 977 | LINC00977 | 792.9kb | Sense | Chr. 8 - 129216467 - 129241240 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000517583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000517583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000517583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000517583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000518044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000518044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000518044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000518044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000518044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000520824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000517583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000517583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000517583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000517583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000518044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000518044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000518044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000518044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000518044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000520824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | ENST00000523173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011796.hg.1 | NR_121673.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | ENST00000520206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | ENST00000520766.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | NR_121672.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800180082.hg.1 | TC0800011796.hg.1 | Clariom D TC |

LINC00824
Genomic Map