| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000517475.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000517475.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000517475.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000517475.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000517475.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000520894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000520894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
ENST00000520894.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000517475.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000517475.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000517475.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000517475.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000517475.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000520894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000520894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
ENST00000520894.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010808.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173405.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173405.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173405.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173405.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173405.hg.1 |
ENST00000514599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800173405.hg.1 |
NR_125388.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800173405.hg.1 |
TC0800010808.hg.1 |
Clariom D TC |