Assay Details
| Gene Symbol: GATA2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125398.1 | 2 - 3 | 760 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DNAJB8 antisense RNA 1 | DNAJB8-AS1 | -17.5kb | Sense | Chr. 3 - 128463594 - 128472317 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC90246 | LOC90246 | 4.6kb | Sense | Chr. 3 - 128507835 - 128510595 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

GATA2-AS1
Genomic Map