| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178230.hg.1 |
TC0800011498.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800178229.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178229.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178229.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178229.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178229.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178229.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178229.hg.1 |
TC0800011498.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178228.hg.1 |
TC0800011498.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800178227.hg.1 |
TC0800011498.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000523557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
ENST00000524283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125416.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011498.hg.1 |
NR_125417.1 |
Refseq |