| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000435967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000458624.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
ENST00000606016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006688.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700146286.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700146286.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700146286.hg.1 |
ENST00000424460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700146286.hg.1 |
NR_125740.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700146286.hg.1 |
TC0700006688.hg.1 |
Clariom D TC |