| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006527.hg.1 |
ENST00000443719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006527.hg.1 |
ENST00000443719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006527.hg.1 |
NR_125745.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006527.hg.1 |
NR_125745.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006527.hg.1 |
ENST00000443719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006527.hg.1 |
ENST00000443719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006527.hg.1 |
NR_125745.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006527.hg.1 |
NR_125745.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144545.hg.1 |
ENST00000443719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144545.hg.1 |
NR_125745.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144545.hg.1 |
ENST00000443719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144545.hg.1 |
NR_125745.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144545.hg.1 |
TC1300006527.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300144546.hg.1 |
ENST00000443719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144546.hg.1 |
NR_125745.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144546.hg.1 |
ENST00000443719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300144546.hg.1 |
NR_125745.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300144546.hg.1 |
TC1300006527.hg.1 |
Clariom D TC |