| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011974.hg.1 |
ENST00000502827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011974.hg.1 |
ENST00000502827.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011974.hg.1 |
NR_125748.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011974.hg.1 |
NR_125748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500182459.hg.1 |
ENST00000502827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500182459.hg.1 |
NR_125748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500182459.hg.1 |
ENST00000502827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500182459.hg.1 |
NR_125748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500182459.hg.1 |
TC0500011974.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500011974.hg.1 |
ENST00000502827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011974.hg.1 |
ENST00000502827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011974.hg.1 |
NR_125748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011974.hg.1 |
NR_125748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500182458.hg.1 |
ENST00000502827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500182458.hg.1 |
NR_125748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500182458.hg.1 |
ENST00000502827.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500182458.hg.1 |
NR_125748.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500182458.hg.1 |
TC0500011974.hg.1 |
Clariom D TC |