Assay Details
| Gene Symbol: LOC101928231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125852.1 | 4 - 5 | 994 | 91 | ||
| GenBank mRNA | BC040308.1 | - | 4 - 5 | 996 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1010 | LINC01010 | Anti-sense | Chr. 6 - 134437716 - 134504020 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC101928304 | LOC101928304 | 77.3kb | Sense | Chr. 6 - 134525318 - 134540005 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928231
Genomic Map