Assay Details
| Gene Symbol: EVADR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125857.1 | 3 - 4 | 475 | 74 | ||
| GenBank mRNA | BU620503.1 | - | 1 - 2 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | collagen type IX alpha 1 | COL9A1 |
DJ149L1.1.2, EDM6, MED, STL4 |
-92.1kb | Anti-sense | Chr. 6 - 70216040 - 70303083 on NCBI Build GRCh38 |
| Downstream | family with sequence similarity 135 member A | FAM135A | KIAA1411 | 14.0kb | Sense | Chr. 6 - 70413404 - 70561174 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

EVADR
Genomic Map