Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009586.hg.1 |
ENST00000440397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009586.hg.1 |
ENST00000440397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009586.hg.1 |
ENST00000440397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009586.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009586.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009586.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009586.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009586.hg.1 |
ENST00000440397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009586.hg.1 |
ENST00000440397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009586.hg.1 |
ENST00000440397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009586.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009586.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009586.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009586.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165715.hg.1 |
ENST00000440397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165715.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165715.hg.1 |
ENST00000440397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165715.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165715.hg.1 |
TC0600009586.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600165717.hg.1 |
ENST00000440397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165717.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165717.hg.1 |
ENST00000440397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165717.hg.1 |
NR_125867.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165717.hg.1 |
TC0600009586.hg.1 |
Clariom D TC |