Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010045.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010045.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010045.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010045.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010045.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010045.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010045.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010045.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010045.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010045.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010045.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010045.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010045.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010045.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010045.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010045.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168605.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168605.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168605.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168605.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168605.hg.1 |
TC0600010045.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600168606.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168606.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168606.hg.1 |
ENST00000429657.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168606.hg.1 |
NR_125873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168606.hg.1 |
TC0600010045.hg.1 |
Clariom D TC |