Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170802.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170802.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170802.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170802.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170802.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170802.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170802.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170802.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170802.hg.1 |
TC0400010380.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400170803.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170803.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170803.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170803.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170803.hg.1 |
TC0400010380.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400170800.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170800.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170800.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170800.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170800.hg.1 |
TC0400010380.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400170801.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170801.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170801.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170801.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170801.hg.1 |
TC0400010380.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400170804.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170804.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170804.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170804.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170804.hg.1 |
TC0400010380.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400170805.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170805.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170805.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170805.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170805.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170805.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170805.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400170805.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400170805.hg.1 |
TC0400010380.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000505018.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
ENST00000512581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010380.hg.1 |
NR_125902.1 |
Refseq |