Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000504955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000504955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000504955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000504955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011519.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000504955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000504955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000504955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000504955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011519.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
ENST00000504955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
ENST00000504955.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179054.hg.1 |
TC0400011519.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400179052.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179052.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179052.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179052.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179052.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179052.hg.1 |
ENST00000506527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179052.hg.1 |
ENST00000509003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179052.hg.1 |
NR_125927.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179052.hg.1 |
TC0400011519.hg.1 |
Clariom D TC |