Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
ENST00000454831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
ENST00000454831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
ENST00000454831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
ENST00000454831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014719.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209583.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209583.hg.1 |
ENST00000454831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209583.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209583.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209583.hg.1 |
ENST00000454831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209583.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209583.hg.1 |
TC0100014719.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
ENST00000454831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
ENST00000454831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
ENST00000454831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
ENST00000454831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014719.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209580.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209580.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209580.hg.1 |
ENST00000443939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209580.hg.1 |
NR_125943.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209580.hg.1 |
TC0100014719.hg.1 |
Clariom D TC |