| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000423175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000423175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000423175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000423175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000423175.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000423988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000423988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000423988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000423988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000451766.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000451766.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000451766.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000451766.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000451766.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000458044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000458044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000458044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
ENST00000458044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234301.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234301.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234301.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234301.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234301.hg.1 |
TC0100017808.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000418557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000423175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000423175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000423175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000423175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000423175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000423988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000423988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000423988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000423988.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000451766.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000451766.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000451766.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000451766.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000451766.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000458044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000458044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000458044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
ENST00000458044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017808.hg.1 |
NR_125945.1 |
Refseq |